Fmoc-d-pipecolic acid ≥96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-pipecolic acid, (R)-1-Fmoc-2-piperidinecarboxylic acid, Fmoc-D-Pip-OH, 1-Fmoc-D-pipecolic acid, Fmoc-D-Homopro-OH, (R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
Purity: 96+%
CAS No.: 101555-63-9
Molecular Formula: C21H21NO4
Formula Weight: 351.40
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 154-159°
Optical Rotation: +25° (c=1 in DMF)
MDL No.: MFCD00235899
Formula:
C₂₁H₂₁NO₄ MW: 351,402 g/mol Melting Pt: 154…159 °C |
MDL Number:
MFCD00235899 CAS Number: 101555-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...