Methyl-2-(3-oxetanylidene)acetate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 2-(oxetan-3-ylidene)acetate
H57376.MD
H57376.ME
H57376.MDEA
134
GBP
H57376.MD
H57376.ME
Methyl-2-(3-oxetanylidene)acetate 95%
Methyl-2-(3-oxetanylidene)acetate
Purity: 95%
CAS No.: 1105665-34-6
Molecular Formula: C6H8O3
Formula Weight: 128.13
MDL No.: MFCD16140399
Formula:
C₆H₈O₃ MW: 128,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16140399 CAS Number: 1105665-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...