2-Chloro-4-iodonicotinic acid methyl ester 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H57319.MD
H57319.03
H57319.MDEA
0
GBP
H57319.MD
H57319.03
2-Chloro-4-iodonicotinic acid methyl ester 97%
2-Chloro-4-iodonicotinic acid methyl ester
Purity: 97%
CAS No.: 185041-05-8
Molecular Formula: C7H5ClINO2
Formula Weight: 297.48
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
Melting Point: 81-85°
MDL No.: MFCD04039871
Formula:
C₇H₅ClINO₂ Melting Pt: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039871 CAS Number: 185041-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...