Pyrrolidin-3-ylmethanol ≥97%

Supplier: Thermo Scientific
Warning

H57225.MD H57225.03
H57225.MDEA 98.8 GBP
H57225.MD H57225.03
Pyrrolidin-3-ylmethanol ≥97%
Pyrrolidin-3-ylmethanol

Purity: 97+% CAS No.: 5082-74-6 Molecular Formula: C5H11NO Formula Weight: 101.15 Storage Temperature: 2°C to 8°C Boiling Point: 165-171° Beilstein Reference: 1420904 MDL No.: MFCD07773080

Formula: C5H11NO
MW: 101,15 g/mol
Boiling Pt: 165…171 °C
Density: 0,979
MDL Number: MFCD07773080
CAS Number: 5082-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR