4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde 95%

Supplier: Thermo Scientific
Warning

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carbaldehyde

H57011.MD H57011.03
H57011.MDEA 30.9 GBP
H57011.MD H57011.03
4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde 95%
4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde

Purity: 95% CAS No.: 343604-24-0 Molecular Formula: C14H9F3O Formula Weight: 250.22 MDL No.: MFCD01631879

Formula: C₁₄H₉F₃O
MW: 250,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631879
CAS Number: 343604-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR