(2-Bromoethoxy)-tert-butyldimethylsilane 98% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with sodium carbonate (Na₂CO₃)

H56594.03 H56594.06
H56594.03EA 0 GBP
H56594.03 H56594.06
(2-Bromoethoxy)-tert-butyldimethylsilane 98% stabilised

Purity: 98% CAS No.: 86864-60-0 Molecular Formula: C8H19BrOSi Formula Weight: 239.23 Flash Point: 73°(163°F) Physical Form: Liquid Density: 1.115 Refractive Index: 1.444 Appearance: Colourless Boiling Point: 120°/100mm MDL No.: MFCD00209550

Formula: C₈H₁₉BrOSi
Boiling Pt: 120 °C (100 mmHg)
Density: 1,115
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00209550
CAS Number: 86864-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR