Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) ≥97%
Supplier: Thermo Fisher Scientific
Synonyms:
[1,2-Bis(diphenylphosphino)ethane]dichlorocobalt(II)
H55020.03
H55020.06
H55020.03EA
0
GBP
H55020.03
H55020.06
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II) ≥97%
Dichloro[bis(1,2-diphenylphosphino)ethane]cobalt(II)
Purity: 97%
CAS No.: 18498-01-6
Molecular Formula: C26H24Cl2CoP2
Formula Weight: 528.26
Melting Point: 260° dec.
MDL No.: MFCD00134173
Formula:
C₂₆H₂₄Cl₂CoP₂ Melting Pt: 260 °C (dec.) |
MDL Number:
MFCD00134173 CAS Number: 18498-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...