(1H-Indol-4-ylmethyl)amine 97%

Supplier: Thermo Fisher Scientific
Warning

H54963.MD H54963.03
H54963.MDEA 36.7 GBP
H54963.MD H54963.03
(1H-Indol-4-ylmethyl)amine 97%
(1H-Indol-4-ylmethyl)amine

Purity: 97% CAS No.: 3468-18-6 Molecular Formula: C9H10N2 Formula Weight: 146.19 Melting Point: 130-135° MDL No.: MFCD04973297

Formula: C₉H₁₀N₂
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD04973297
CAS Number: 3468-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR