Phenanthrene-4,5-dicarboxylic acid 97%

Supplier: Thermo Scientific
Warning

H54939.MD H54939.03
H54939.MDEA 52 GBP
H54939.MD H54939.03
Phenanthrene-4,5-dicarboxylic acid 97%
Phenanthrene-4,5-dicarboxylic acid

Purity: 97% CAS No.: 5462-82-8 Molecular Formula: C16H10O4 Formula Weight: 266.25 Melting Point: 244-246° MDL No.: MFCD11110581

Formula: C₁₆H₁₀O₄
MW: 266,25 g/mol
Melting Pt: 244…246 °C
Storage Temperature: Ambient
MDL Number: MFCD11110581
CAS Number: 5462-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR