5-Bromo-2-methylpyridin-3-amine 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Amino-5-bromo-2-methylpyridine
H54665.MD
H54665.03
H54665.MDEA
108
GBP
H54665.MD
H54665.03
5-Bromo-2-methylpyridin-3-amine 96%
5-Bromo-2-methylpyridin-3-amine
Purity: 96%
CAS No.: 914358-73-9
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Melting Point: 108-109°
MDL No.: MFCD09031418
Formula:
C₆H₇BrN₂ MW: 187,04 g/mol Melting Pt: 108…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD09031418 CAS Number: 914358-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...