2-(3-Methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methyl-2-(3-methoxyphenyl)thiazole-5-carboxylic acid

H54566.MD H54566.03 H54566.06
H54566.MDEA 29.6 GBP
H54566.MD H54566.03 H54566.06
2-(3-Methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid 97%
2-(3-Methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid

Purity: 97% CAS No.: 879636-95-0 Molecular Formula: C12H11NO3S Formula Weight: 249.28 MDL No.: MFCD06857927

Formula: C₁₂H₁₁NO₃S
MW: 249,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06857927
CAS Number: 879636-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR