6-Bromo-4-hydroxyquinoline-3-carboxylic acid 96%
Supplier: Thermo Scientific
|
Warning
|
H54441.03
H54441.06
H54441.03EA
113
GBP
H54441.03
H54441.06
6-Bromo-4-hydroxyquinoline-3-carboxylic acid 96%
6-Bromo-4-hydroxyquinoline-3-carboxylic acid
Purity: 96%
CAS No.: 98948-95-9
Molecular Formula: C10H6BrNO3
Formula Weight: 268.06
MDL No.: MFCD01569278
Formula:
C₁₀H₆BrNO₃ MW: 268,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01569278 CAS Number: 98948-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...