5-Bromo-N,3-dimethyl-2-pyridinamine 96%

Supplier: Thermo Scientific
Warning

Synonyms: (5-Bromo-3-methylpyridin-2-yl)methylamine, 5-Bromo-3-methyl-2-(methylamino)pyridine, 5-Bromo-N,3-dimethylpyridin-2-amine

H54360.MD H54360.03
H54360.MDEA 106 GBP
H54360.MD H54360.03
5-Bromo-N,3-dimethyl-2-pyridinamine 96%
5-Bromo-N,3-dimethyl-2-pyridinamine

Purity: 96% CAS No.: 245765-66-6 Molecular Formula: C7H9BrN2 Formula Weight: 201.06 Storage Temperature: 2°C to 8°C Melting Point: 62-64° MDL No.: MFCD10000482

Formula: C₇H₉BrN₂
MW: 201,07 g/mol
Melting Pt: 62…64 °C
MDL Number: MFCD10000482
CAS Number: 245765-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR