3-(Di-n-propylsulphamoyl)benzeneboronic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H54354.MD
H54354.03
H54354.MDEA
0
GBP
H54354.MD
H54354.03
3-(Di-n-propylsulphamoyl)benzeneboronic acid 97%
3-(Di-n-propylsulphamoyl)benzeneboronic acid
Purity: 97%
CAS No.: 1449145-31-6
Molecular Formula: C12H20BNO4S
Formula Weight: 285.17
MDL No.: MFCD20265249
Formula:
C₁₂H₂₀BNO₄S Storage Temperature: Ambient |
MDL Number:
MFCD20265249 CAS Number: 1449145-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...