2-Amino-5-bromo-4-methylpyrimidine 96%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Amino-4-methyl-5-bromopyrimidine

H54186.03 H54186.06
H54186.03EA 43.5 GBP
H54186.03 H54186.06
2-Amino-5-bromo-4-methylpyrimidine 96%
2-Amino-5-bromo-4-methylpyrimidine

Purity: 96% CAS No.: 17321-93-6 Molecular Formula: C5H6BrN3 Formula Weight: 188.03 Storage Temperature: 2°C to 8°C Melting Point: 192-194° MDL No.: MFCD00094469

Formula: C₅H₆BrN₃
MW: 188,03 g/mol
Melting Pt: 192…194 °C
MDL Number: MFCD00094469
CAS Number: 17321-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR