O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,1,3,3-Tetramethyl-2-(4-oxobenzo[d][1,2,3]triazin-3(4H)-yl)isouronium tetrafluoroborate, N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium Tetrafluoroborate, TDBTU
H54144.03
H54144.06
H54144.03EA
38.7
GBP
H54144.03
H54144.06
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98%
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate
Purity: 98+%
CAS No.: 125700-69-8
Molecular Formula: C12H16BF4N5O2
Formula Weight: 349.09
Storage Temperature: 2°C to 8°C
MDL No.: MFCD00077730
Formula:
C₁₂H₁₆BF₄N₅O₂ MW: 349,10 g/mol |
MDL Number:
MFCD00077730 CAS Number: 125700-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...