2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%

Supplier: Thermo Scientific
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine-2-carboxylic acid methylamide, N-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

H54084.MD H54084.03
H54084.MDEA 108 GBP
H54084.MD H54084.03
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
2-(Methylcarbamoyl)pyridine-5-boronic acid pinacol ester

Purity: 96% CAS No.: 945863-21-8 Molecular Formula: C13H19BN2O3 Formula Weight: 262.11 Storage Temperature: 2°C to 8°C Melting Point: 113-115° MDL No.: MFCD08458483

Formula: C₁₃H₁₉BN₂O₃
MW: 262,11 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD08458483
CAS Number: 945863-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR