4-(Trifluoromethyl)hydrocinnamic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-[4-(Trifluoromethyl)phenyl]propanoic acid

H54010.03 H54010.06
H54010.03EA 42.4 GBP
H54010.03 H54010.06
4-(Trifluoromethyl)hydrocinnamic acid 97%
4-(Trifluoromethyl)hydrocinnamic acid

Purity: 97% CAS No.: 53473-36-2 Molecular Formula: C10H9F3O2 Formula Weight: 218.17 Melting Point: 99° MDL No.: MFCD00674032

Formula: C₁₀H₉F₃O₂
MW: 218,17 g/mol
Melting Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00674032
CAS Number: 53473-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR