5-Norbornene-2-exo,3-exo-dimethanol 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-exo,6-exo-Bis(hydroxymethyl)-2-norbornene, Bicyclo[2.2.1]hept-5-ene-2-exo,3-exo-dimethanol
H53422.03
H53422.06
H53422.14
H53422.03EA
76.9
GBP
H53422.03
H53422.06
H53422.14
5-Norbornene-2-exo,3-exo-dimethanol 97%
5-Norbornene-2-exo,3-exo-dimethanol
Purity: 97%
CAS No.: 699-95-6
Molecular Formula: C9H14O2
Formula Weight: 154.21
Flash Point: 113°(235°F)
Refractive Index: 1.523
Boiling Point: 97°/20mm
MDL No.: MFCD00799568
Formula:
C₉H₁₄O₂ MW: 154,21 g/mol Boiling Pt: 97 °C (20 mmHg) Density: 1,027 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00799568 CAS Number: 699-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...