Boc-(S)-2-amino-3-ethoxypropionic acid 97%

Supplier: Thermo Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-ethoxypropanoic acid

H52781.MD H52781.03
H52781.MDEA 131 GBP
H52781.MD H52781.03
Boc-(S)-2-amino-3-ethoxypropionic acid 97%
Boc-(S)-2-amino-3-ethoxypropionic acid

Purity: 97% CAS No.: 104839-00-1 Molecular Formula: C10H19NO5 Formula Weight: 233.26 MDL No.: MFCD06797558

Formula: C₁₀H₁₉NO₅
MW: 233,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797558
CAS Number: 104839-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR