(R)-2-Methylpyrrolidine-p-toluenesulphonate 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H52410.03
H52410.06
H52410.03EA
0
GBP
H52410.03
H52410.06
(R)-2-Methylpyrrolidine-p-toluenesulphonate 97%
(R)-2-Methylpyrrolidine-p-toluenesulphonate
Purity: 97%
CAS No.: 204387-55-3
Molecular Formula: C12H19NO3S
Formula Weight: 257.35
MDL No.: MFCD07368911
Formula:
C₁₂H₁₉NO₃S MW: 257.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368911 CAS Number: 204387-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...