5-Amino-2-cyano-3-methoxypyridine 96%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Amino-3-methoxypicolinonitrile

H52352.MD H52352.03
H52352.MDEA 125 GBP
H52352.MD H52352.03
5-Amino-2-cyano-3-methoxypyridine 96%
5-Amino-2-cyano-3-methoxypyridine

Purity: 96% CAS No.: 573762-39-7 Molecular Formula: C7H7N3O Formula Weight: 149.15 MDL No.: MFCD11111246

Formula: C₇H₇N₃O
MW: 149,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111246
CAS Number: 573762-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR