(S)-1,4-Diazabicyclo[4.3.0]nonane ≥98%
Supplier: Thermo Scientific
|
Danger
|
H52295.MD
H52295.03
H52295.MDEA
192
GBP
H52295.MD
H52295.03
(S)-1,4-Diazabicyclo[4.3.0]nonane ≥98%
(S)-1,4-Diazabicyclo[4.3.0]nonane
Purity: 98+%
CAS No.: 93643-24-4
Molecular Formula: C7H14N2
Formula Weight: 126.20
UN No.: UN3267
MDL No.: MFCD03787926
Formula:
C₇H₁₄N₂ MW: 126,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03787926 CAS Number: 93643-24-4 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...