4,5-Diphenyl-1,3-thiazol-2-amine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4,5-Diphenylthiazol-2-amine
H51818.03
H51818.06
H51818.03EA
41.5
GBP
H51818.03
H51818.06
4,5-Diphenyl-1,3-thiazol-2-amine 97%
4,5-Diphenyl-1,3-thiazol-2-amine
Purity: 97%
CAS No.: 6318-74-7
Molecular Formula: C15H12N2S
Formula Weight: 252.33
Odour: Odourless
MDL No.: MFCD00438803
Formula:
C₁₅H₁₂N₂S MW: 252,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00438803 CAS Number: 6318-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...