4-Amino-1-(2-pyridyl)piperidine hydrochloride 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H51108.MD
H51108.03
H51108.MDEA
50.9
GBP
H51108.MD
H51108.03
4-Amino-1-(2-pyridyl)piperidine hydrochloride 97%
4-Amino-1-(2-pyridyl)piperidine hydrochloride
Purity: 97%
CAS No.: 77145-39-2
Molecular Formula: C10H15N3.HCl
Formula Weight: 213.71
MDL No.: MFCD09607833
Formula:
C₁₀H₁₆ClN₃ MW: 213.709 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09607833 CAS Number: 77145-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...