1-(3-Methylpyridin-2-yl)-1,4-diazepane 95%

Supplier: Thermo Scientific
Danger

H51006.03 H51006.MD
H51006.03EA 233 GBP
H51006.03 H51006.MD
1-(3-Methylpyridin-2-yl)-1,4-diazepane 95%
1-(3-Methylpyridin-2-yl)-1,4-diazepane

Purity: 95% CAS No.: 880362-05-0 Molecular Formula: C11H17N3 Formula Weight: 191.28 Refractive Index: 1.5735 UN No.: UN2735 MDL No.: MFCD08061111

Formula: C₁₁H₁₇N₃
Storage Temperature: Ambient
MDL Number: MFCD08061111
CAS Number: 880362-05-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR