4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine 97%
Supplier: Thermo Scientific
|
Warning
|
H50873.03
H50873.MD
H50873.03EA
191
GBP
H50873.03
H50873.MD
4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine 97%
4-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine
Purity: 97%
CAS No.: 733748-92-0
Molecular Formula: C10H17N3O
Formula Weight: 195.27
Refractive Index: 1.4860
MDL No.: MFCD08061038
Formula:
C₁₀H₁₇N₃O Storage Temperature: Ambient |
MDL Number:
MFCD08061038 CAS Number: 733748-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...