2-(4-Piperidinyloxy)-N-propylacetamide 95%

Supplier: Thermo Scientific
Warning

H50865.03 H50865.MD
H50865.03EA 253 GBP
H50865.03 H50865.MD
2-(4-Piperidinyloxy)-N-propylacetamide 95%
2-(4-Piperidinyloxy)-N-propylacetamide

Purity: 95% CAS No.: 902836-22-0 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD08064265

Formula: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064265
CAS Number: 902836-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR