α-3-Dimethylstyrene
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Methyl-3-(prop-1-en-2-yl)benzene
H50760.03
H50760.MD
H50760.03EA
282
GBP
H50760.03
H50760.MD
α-3-Dimethylstyrene
alpha-3-Dimethylstyrene
CAS No.: 1124-20-5
Molecular Formula: C10H12
Formula Weight: 132.21
Storage Temperature: 2°C to 8°C
Odour: Characteristic
UN No.: UN3295
MDL No.: MFCD08060972
Formula:
C₁₀H₁₂ MW: 132,21 g/mol |
MDL Number:
MFCD08060972 CAS Number: 1124-20-5 EINECS: 214-394-9 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...