2-Phenoxypropylamine 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
H50720.03
H50720.MD
H50720.03EA
273
GBP
H50720.03
H50720.MD
2-Phenoxypropylamine 98%
2-Phenoxypropylamine
Purity: 98%
CAS No.: 6437-49-6
Molecular Formula: C9H13NO
Formula Weight: 151.21
UN No.: UN3259
MDL No.: MFCD07781046
Formula:
C₉H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD07781046 CAS Number: 6437-49-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...