2-Ethoxy-4-formylphenyl acetate 99%

Supplier: Thermo Scientific
Warning

Synonyms: Ethylvanillin acetate

H50488.03 H50488.MD
H50488.03EA 253 GBP
H50488.03 H50488.MD
2-Ethoxy-4-formylphenyl acetate 99%
2-Ethoxy-4-formylphenyl acetate

Purity: 99% CAS No.: 72207-94-4 Molecular Formula: C11H12O4 Formula Weight: 208.22 MDL No.: MFCD01666413

Formula: C₁₁H₁₂O₄
MW: 208,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01666413
CAS Number: 72207-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR