1-Naphthylacetyl chloride 95%

Supplier: Thermo Scientific
Danger

H50386.03 H50386.06
H50386.03EA 88.8 GBP
H50386.03 H50386.06
1-Naphthylacetyl chloride 95%
1-Naphthylacetyl chloride

Purity: 95% CAS No.: 5121-00-6 Molecular Formula: C12H9ClO Formula Weight: 204.66 Refractive Index: 1.6210 Odour: Pungent Boiling Point: 130°/0.6mm UN No.: UN3265 MDL No.: MFCD00175852

Formula: C₁₂H₉ClO
MW: 204,66 g/mol
Boiling Pt: 130 °C (0,6 mmHg)
Density: 1,23
Storage Temperature: Ambient
MDL Number: MFCD00175852
CAS Number: 5121-00-6
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR