2-Phthalimidoethanesulphonyl chloride 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(1,3-Dioxoisoindolin-2-yl)ethanesulphonylchloride
H50382.03
H50382.06
H50382.03EA
82.4
GBP
H50382.03
H50382.06
2-Phthalimidoethanesulphonyl chloride 97%
2-Phthalimidoethanesulphonyl chloride
Purity: 97%
CAS No.: 4403-36-5
Molecular Formula: C10H8ClNO4S
Formula Weight: 273.70
Melting Point: 156-160°
UN No.: UN3261
MDL No.: MFCD01861217
Formula:
C₁₀H₈ClNO₄S MW: 273,7 g/mol Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861217 CAS Number: 4403-36-5 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...