Palladium 5% on carbon paste wetted with ca. 50% water A405028-5
Supplier: Thermo Scientific
H36996.06
H36996.14
H36996.22
H36996.06EA
0
GBP
H36996.06
H36996.14
H36996.22
Palladium 5% on carbon paste wetted with ca. 50% water A405028-5
Palladium
CAS No.: 7440-05-3
Physical Form: Solid
Appearance: Dark grey
Odour: Odourless
MDL No.: MFCD03457879
Formula:
Pd MW: 106,420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...