Rhodium 5% on alumina powder C301099-5

Supplier: Thermo Scientific

H36201.06 H36201.14 H36201.22
H36201.06EA 665 GBP
H36201.06 H36201.14 H36201.22
Rhodium 5% on alumina powder C301099-5
Rhodium

CAS No.: 7440-16-6 Physical Form: Powder Appearance: Grey Odour: Odourless MDL No.: MFCD00011201

Formula: Rh
MW: 102,9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR