2-Ethoxyacetyl chloride 97% stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilisation: Stabiised with magnesium oxide (MgO) 0,3%

H34443.06 H34443.14 H34443.22
H34443.06EA 62.8 GBP
H34443.06 H34443.14 H34443.22
2-Ethoxyacetyl chloride 97% stabilised
2-Ethoxyacetyl chloride

Stab. with 0.3% Magnesium oxide


Purity: 97% CAS No.: 14077-58-8 Molecular Formula: C4H7ClO2 Formula Weight: 122.55 UN No.: UN2920 MDL No.: MFCD11170566

Formula: C₄H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11170566
CAS Number: 14077-58-8
EINECS: 237-930-3
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR