8-Methoxyjulolidine 95%

Supplier: Thermo Fisher Scientific
Warning

H34100.03 H34100.09
H34100.03EA 18.8 GBP
H34100.03 H34100.09
8-Methoxyjulolidine 95%
8-Methoxyjulolidine

Purity: 95% CAS No.: 63468-83-7 Molecular Formula: C13H17NO Formula Weight: 203.28 MDL No.: MFCD00037096

Formula: C₁₃H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD00037096
CAS Number: 63468-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR