2-Mercapto-3-phenyl-3,5,6,7-tetrahydro-4H-cyclopenta[b]thieno[2,3-d]pyrimidin-4-one 96%

Supplier: Thermo Fisher Scientific
Warning

H33881.MD H33881.03 H33881.06
H33881.MDEA 0 GBP
H33881.MD H33881.03 H33881.06
2-Mercapto-3-phenyl-3,5,6,7-tetrahydro-4H-cyclopenta[b]thieno[2,3-d]pyrimidin-4-one 96%
2-Mercapto-3-phenyl-3,5,6,7-tetrahydro-4H-cyclopenta[b]thieno[2,3-d]pyrimidin-4-one

Purity: 96% CAS No.: 300557-77-1 Molecular Formula: C15H12N2OS2 Formula Weight: 300.40 Melting Point: 250° MDL No.: MFCD01018198

Formula: C₁₅H₁₂N₂OS₂
MW: 300.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01018198
CAS Number: 300557-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR