Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 96%

Supplier: Thermo Scientific
Warning

Synonyms: Ethyl 2-amino-6-methyl-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

H33760.03 H33760.06
H33760.03EA 39.1 GBP
H33760.03 H33760.06
Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate 96%
Ethyl-2-amino-6-methyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Purity: 96% CAS No.: 76981-71-0 Molecular Formula: C12H17NO2S Formula Weight: 239.34 Melting Point: 110-112° MDL No.: MFCD00638882

Formula: C₁₂H₁₇NO₂S
MW: 239,34 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00638882
CAS Number: 76981-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR