2-(2-Methylphenyl)thioacetamide 97%

Supplier: Thermo Scientific
Danger

H33111.03 H33111.06
H33111.03EA 49.8 GBP
H33111.03 H33111.06
2-(2-Methylphenyl)thioacetamide 97%
2-(2-Methylphenyl)thioacetamide

Purity: 97% CAS No.: 34243-66-8 Molecular Formula: C9H11NS Formula Weight: 165.26 Melting Point: 120-124° MDL No.: MFCD09757579

Formula: C₉H₁₁NS
MW: 165,26 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09757579
CAS Number: 34243-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR