Agarose 6% B, beads
Supplier: Thermo Scientific
|
Warning
|
H32491.AUEA
528
GBP
H32491.AU
Agarose 6% B, beads
Agarose
CAS No.: 9012-36-6
Storage Temperature: 2°C to 8°C
Flash Point: 38°(100°F)
Boiling Point: 100°
UN No.: UN1170
MDL No.: MFCD00081294
Boiling Pt:
100 °C Density: 1,040 Flash Pt: 38 °C (100 °F) |
MDL Number:
MFCD00081294 CAS Number: 9012-36-6 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...