2-Bromo-3-fluoropropionic acid 96%
Supplier: Thermo Scientific
|
Danger
|
H31718.MD
H31718.03
H31718.MDEA
90.8
GBP
H31718.MD
H31718.03
2-Bromo-3-fluoropropionic acid 96%
2-Bromo-3-fluoropropionic acid
Purity: 96%
Molecular Formula: C3H4BrFO2
Formula Weight: 170.97
Boiling Point: 62-63°/0.2mm
UN No.: UN3265
MDL No.: MFCD09800640
Formula:
C₃H₄BrFO₂ Boiling Pt: 62…63 °C (0,2 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD09800640 CAS Number: 39621-37-9 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...