4-Benzyloxy-2-fluorobenzoic acid 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-(Benzyloxy)-2-fluorobenzoic acid
H31488.03
H31488.06
H31488.03EA
19.1
GBP
H31488.03
H31488.06
4-Benzyloxy-2-fluorobenzoic acid 98%
4-Benzyloxy-2-fluorobenzoic acid
Purity: 98%
CAS No.: 114045-96-4
Molecular Formula: C14H11FO3
Formula Weight: 246.24
Melting Point: 165-168°
MDL No.: MFCD12407082
Formula:
C₁₄H₁₁FO₃ Melting Pt: 165…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD12407082 CAS Number: 114045-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...