Vinyl decanoate 95% stabilised
Supplier: Thermo Scientific
Synonyms:
Decanoic Acid Vinyl Ester
H29081.14
H29081.06
H29081.14EA
90.2
GBP
H29081.14
H29081.06
Vinyl decanoate 95% stabilised
Vinyl decanoate
Purity: 95%
CAS No.: 4704-31-8
Molecular Formula: C12H22O
Formula Weight: 198.30
Flash Point: 104°(219°F)
Refractive Index: 1.435
Boiling Point: 119-120°/10mm
MDL No.: MFCD00027349
Formula:
C₁₂H₂₂O₂ MW: 198,31 g/mol Boiling Pt: 119…120 °C (10 mmHg) Density: 0,886 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027349 CAS Number: 4704-31-8 EINECS: 225-185-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...