3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester 97%

Supplier: Thermo Fisher Scientific

H28599.06 H28599.03
H28599.06EA 0 GBP
H28599.06 H28599.03
3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester 97%
3-(Furfurylaminocarbonyl)benzeneboronic acid pinacol ester

Purity: 97% CAS No.: 1073353-63-5 Molecular Formula: C18H22BNO4 Formula Weight: 327.18 Melting Point: 122-124° MDL No.: MFCD09266173

Formula: C₁₈H₂₂BNO₄
MW: 327.18 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09266173
CAS Number: 1073353-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR