4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥95%
Supplier: Thermo Scientific
Synonyms:
1-Methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine, 4-(4-Methylpiperazino)methylphenylboronic acid, pinacol ester
H28495.06
H28495.03
H28495.06EA
0
GBP
H28495.06
H28495.03
4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥95%
4-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Purity: 95%
CAS No.: 938043-30-2
Molecular Formula: C18H29BN2O2
Formula Weight: 316.25
Melting Point: 61-65°
MDL No.: MFCD09266182
Formula:
C₁₈H₂₉BN₂O₂ MW: 316,2461 g/mol Melting Pt: 61…65 °C |
MDL Number:
MFCD09266182 CAS Number: 938043-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...