(4-Isopropylphenoxy)acetyl chloride 98%
Supplier: Thermo Scientific
|
Danger
|
H28036.06
H28036.03
H28036.06EA
89.4
GBP
H28036.06
H28036.03
(4-Isopropylphenoxy)acetyl chloride 98%
(4-Isopropylphenoxy)acetyl chloride
Purity: 98%
CAS No.: 223128-33-4
Molecular Formula: C11H13ClO2
Formula Weight: 212.67
Refractive Index: 1.5195
UN No.: UN3265
MDL No.: MFCD02628421
Formula:
C₁₁H₁₃ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD02628421 CAS Number: 223128-33-4 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...