1-Ethyl-4-[(4-n-propylphenyl)ethynyl]benzene ≥99%

Supplier: Thermo Scientific

H27398.09 H27398.03
H27398.09EA 202 GBP
H27398.09 H27398.03
1-Ethyl-4-[(4-n-propylphenyl)ethynyl]benzene ≥99%
1-Ethyl-4-[(4-n-propylphenyl)ethynyl]benzene

Purity: 99+% CAS No.: 102225-55-8 Molecular Formula: C19H20 Formula Weight: 248.37 Melting Point: 49-51° MDL No.: MFCD05864410

Formula: C₁₉H₂₀
MW: 248,37 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD05864410
CAS Number: 102225-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR