Tris(cyclopentadienyl)erbium(III) 99% (99.9% Er)

Supplier: Thermo Fisher Scientific
Warning

H27389.03 H27389.06
H27389.03EA 0 GBP
H27389.03 H27389.06
Tris(cyclopentadienyl)erbium(III) 99% (99.9% Er)
Tris(cyclopentadienyl)erbium(III)

Purity: 99% CAS No.: 39330-74-0 Molecular Formula: C15H15Er Formula Weight: 362.54 Melting Point: 287° UN No.: UN3181 MDL No.: MFCD00145475

Formula: C₁₅H₁₅Er
Melting Pt: 287 °C
Storage Temperature: Ambient
MDL Number: MFCD00145475
CAS Number: 39330-74-0
EINECS: 254-416-4
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR