Bis(methylcyclopentadienyl)zirconium dichloride 98%

Supplier: Thermo Scientific
Danger

H27241.03 H27241.06
H27241.03EA 73.7 GBP
H27241.03 H27241.06
Bis(methylcyclopentadienyl)zirconium dichloride 98%
Bis(methylcyclopentadienyl)zirconium dichloride

Purity: 98% CAS No.: 12109-71-6 Molecular Formula: C12H14Cl2Zr Formula Weight: 320.37 Melting Point: 180-184° UN No.: UN3261 MDL No.: MFCD00672106

Formula: C₁₂H₁₄Cl₂Zr
MW: 320,37 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00672106
CAS Number: 12109-71-6
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR